Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Regulatory protein GAL4 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP276808SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP276808SS 0.025 mg
NBP276808 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP276808SS Supplier Novus Biologicals Supplier No. NBP276808SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Regulatory protein GAL4 Polyclonal specifically detects Regulatory protein GAL4 in Yeast samples. It is validated for Western Blot.

Specifications

Antigen Regulatory protein GAL4
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1 - 2 ug/ml
Formulation PBS with 0.02% sodium azide
Host Species Rabbit
Immunogen Partial recombinant protein made to a N terminal portion of the yeast GAL4 protein (amino acids 1-147) [UniProt P04386]
Purification Method Affinity Purified
Quantity 0.025 mg
Primary or Secondary Primary
Target Species Yeast
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.