Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BINDER™ Reinforced Shelf
SDP

Catalog No. 08111305
Click to view available options
Material:
Steel

For Use With: KBF P 720

  • Reinforced
  • Positioned at bottom level
  • Max. load 67 kg
  • Includes additional attachment for operation of shaking device, stirring device or roller bottle system

Specifications

Description Shelf, reinforced - positioned at bottom level, max. load 45 kg, with additional attachment for operation of shaking device, stirring device or roller bottle system
For Use With (Equipment) BINDER Chambers KBF P 720
Holds 67 kg
Material Steel
Type Shelf
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.