Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RelA/NFkB p65 Antibody (112A1021) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 13 publications

$244.50 - $562.50

Specifications

Antigen RelA/NFkB p65
Clone 112A1021
Concentration 1.0 mg/mL
Dilution Western Blot 2 - 5 ug/mL. Use reported in scientific literature (Wittmann et al (2005)), Simple Western 1:25. Use reported by customer review, Flow Cytometry 0.25 - 0.5 ug/10^6 cells, Immunohistochemistry 5 - 15 ug/mL, Immunoprecipitation reported by customer review, Immunohistochemistry-Paraffin 5 ug/mL. Use reported in scientific literature (Chang et al (2005)), Flow (Intracellular) reported in scientific literature (Wittmann et al (2005)), CyTOF-ready
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), CyTOF
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056712T
SDP
View Documents
Novus Biologicals
NB10056712SS
0.025 mg
Each for $244.50
Only null left
Add to Cart
 
NB10056712
SDP
View Documents
Novus Biologicals
NB10056712
0.1 mg
Each for $562.50
Only null left
Add to Cart
 
Description

Description

RelA/NFkB p65 Monoclonal specifically detects RelA/NFkB p65 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Flow (Intracellular), CyTOF-ready.
Specifications

Specifications

RelA/NFkB p65
1.0 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), CyTOF
Unconjugated
Mouse
Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Signal Transduction, Transcription Factors and Regulators
Q969V5
5970
This NfkB p65 antibody was raised against a synthetic peptide corresponding to amino acids 526-539 of human NF-kB (p65) protein.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
112A1021
Western Blot 2 - 5 ug/mL. Use reported in scientific literature (Wittmann et al (2005)), Simple Western 1:25. Use reported by customer review, Flow Cytometry 0.25 - 0.5 ug/10^6 cells, Immunohistochemistry 5 - 15 ug/mL, Immunoprecipitation reported by customer review, Immunohistochemistry-Paraffin 5 ug/mL. Use reported in scientific literature (Chang et al (2005)), Flow (Intracellular) reported in scientific literature (Wittmann et al (2005)), CyTOF-ready
Monoclonal
Purified
RUO
Human, Mouse, Rat
anti-NF kB p65, anti-NFkB p65, anti-NF-kB p65, anti-p65, anti-RelA, anti-rela p65, MGC131774, nf kb p65, NFkB p65, NF-kB p65, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65, p65 flow cytometry, p65 immunohistochemistry, RelA, rela p65, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65
RELA
IgG1 κ
Protein G purified
Cross reacts with Human, Mouse and Rat. Predicted to react with Cow, Gorilla, New World Monkey and Pig.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.