Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RelA/NFkB p65 Antibody (465003), Alexa Fluor™ 647, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB50781R
Description
RelA/NFkB p65 Monoclonal antibody specifically detects RelA/NFkB p65 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| RelA/NFkB p65 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 465003 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| MGC131774, nf kb p65, NFkB p65, NF-kB p65, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65, RelA, rela p65, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65 | |
| E. coli-derived recombinant human RelA/NF kappa B p65, Met1-Thr254, Accession # NP_068810 | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| 5970 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction