Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RelA/NFkB p65 Antibody (532301), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB5078X
Description
RelA/NFkB p65 Monoclonal antibody specifically detects RelA/NFkB p65 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF, Gene Knock-OutSpecifications
RelA/NFkB p65 | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF, Gene Knock-Out | |
532301 | |
Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready, Knockout Validated | |
MGC131774, nf kb p65, NFkB p65, NF-kB p65, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65, RelA, rela p65, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65 | |
E. coli-derived recombinant human RelA/NF kappa B p65 isoform 1, Asn456-Ser551, Accession # Q04206 | |
0.1 mL | |
Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Signal Transduction, Transcription Factors and Regulators | |
5970 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction