Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ RelA/NFkB p65 NLS Antibody Blocking Peptide

Click to view available options
Quantity:
0.05 mg
Description
- No Preservative
Specifications
Specifications
Gene ID (Entrez) | 5970 |
Protein Family | RelA/NFkB p65 NLS |
Common Name | RelA/NFkB p65 NLS Blocking Peptide |
Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
For Use With (Application) | Antibody Competition |
Gene Alias | MGC131774, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65 NLS, p65 Nuclear Localization Sequence, p65-NLS, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65 |
Gene Symbol | RELA |
Label Type | Unlabeled |
Product Type | Peptide |
Quantity | 0.05 mg |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction