Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Relaxin 2 Monoclonal Antibody (349102)
GREENER_CHOICE

Catalog No. PIMA524088
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524088 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524088 Supplier Invitrogen™ Supplier No. MA524088
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This antibody detects both the pro and mature forms of human Relaxin-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Relaxin-1 or rhRelaxin-3 is observed. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

Relaxin 1 (also referred to as Relaxin or RLXH1) is a peptide hormone produced by the corpora lutea of the ovary during pregnancy in many mammalian species, including human. The secretion of the hormone into the blood stream just before parturition results in a marked softening and lengthening of the pubic symphysis and a softening of the cervix, which facilitates the birth process. By inhibiting uterine contractions, Relaxin 1 may influence the timing of parturition. Like insulin, Relaxin 1 consists of two peptide chains, A and B, covalently linked by disulfide bonds. By further analogy to insulin, the two peptides are synthesized as a single-chain precursor polypeptide with the B chain at the amino-terminus. The gene that encodes the human Relaxin 1 protein maps to chromosome 9. Relaxin 2, a related protein, is selectively expressed in the ovary during pregnancy. The gene that encodes the human Relaxin 2 protein also maps to chromosome 9.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Relaxin 2
Applications Neutralization, Western Blot
Classification Monoclonal
Clone 349102
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene RLN2
Gene Accession No. P04090
Gene Alias bA12D24.1.1; bA12D24.1.2; H2; H2-preprorelaxin; H2-RLX; Prorelaxin H2; relaxin 2; Relaxin A chain; Relaxin B chain; relaxin H2; relaxin, ovarian, of pregnancy; RLN2; RLX H2-RLX; RLXH2
Gene Symbols RLN2
Host Species Rat
Immunogen E. coli-derived recombinant human Relaxin-2 Val23-Cys185.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6019
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.