Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Relaxin-3 Antibody (MM0526-7G14), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211866AF750
Description
Relaxin-3 Monoclonal specifically detects Relaxin-3 in Human samples. It is validated for Western Blot.Specifications
Relaxin-3 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
RLN3 | |
Human recombinant Relaxin-3 | |
Primary | |
Human | |
IgG2 |
Western Blot | |
MM0526-7G14 | |
Western Blot | |
H3, insl7, relaxin 3, RXN3, ZINS4 | |
Mouse | |
0.1 mL | |
117579 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction