Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Relaxin R2/LGR8 Antibody (1013903) [Janelia Fluor™ 646], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB102501J
Description
Relaxin R2/LGR8 Monoclonal antibody specifically detects Relaxin R2/LGR8 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
Relaxin R2/LGR8 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
1013903 | |
Flow Cytometry, CyTOF-ready | |
G protein coupled receptor affecting testicular descent, GPR106G-protein coupled receptor 106, GREATRelaxin family peptide receptor 2, INSL3R, leucine-rich repeat-containing G protein-coupled receptor 8, Leucine-rich repeat-containing G-protein coupled receptor 8, LGR8.1, LGR8G-protein coupled receptor affecting testicular descent, relaxin receptor 2, relaxin/insulin-like family peptide receptor 2, RXFPR2 | |
Chinese hamster ovary cell line CHO-derived recombinant human Relaxin R2, Met1-Asn416, Accession # Q8WXD0 | |
0.1 mL | |
GPCR | |
122042 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction