Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RelB Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP122980B
Description
RelB Polyclonal antibody specifically detects RelB in Human, Mouse samples. It is validated for Western BlotSpecifications
RelB | |
Polyclonal | |
Western Blot | |
I-Rel, IREL, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, v-rel reticuloendotheliosis viral oncogene homolog B | |
The immunogen recognized by this antibody maps to a region between residue 529 and 579 of human v-rel reticuloendotheliosis viral oncogene homolog B using the numbering given in entry NP_006500.2 (GeneID 5971). | |
0.1 mL | |
Phospho Specific | |
5971 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Biotin | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction