Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega ReliaPrep miRNA Cell and Tissue Miniprep System

Catalog No. p-7112449 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
10 Preps
250 Preps
50 Preps
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRZ6210 10 Preps
PRZ6211 50 Preps
PRZ6212 250 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRZ6210 Supplier Promega Supplier No. Z6210
Only null left
Add to Cart
Add to Cart

ReliaPrep miRNA Cell and Tissue Miniprep System provides complete isolation of total RNA, including microRNA (miRNA) and other small non-coding RNA (sncRNA) subspecies, from a wide variety of cell and tissue types in as quickly as 40 minutes.

  • Fast, Phenol-Free miRNA Isolation from Cells and Tissues
  • Isolate concentrated, quality RNA in only 40 minutes
  • Eliminate harsh organic reagents
  • Work with small samples and low elution volumes

Specifications

Content And Storage 15°C to 30°C
For Use With (Application) Isolation of total RNA
Includes Item Size
Collection Tubes 2 × 10/pk
ReliaPrep™ Minicolumns 2 × 10/pk
1-Thioglycerol 1 × 75μl
DNase Buffer, 10X 1 × 500μl
Nuclease-Free Water 1 × 1,250μl
Lysis Buffer (LBA) 1 × 6mL
Elution Tubes 1 × 25/pk
RNA Wash Solution (RWA) 1 × 11.8mL
DNase I (lyophilized) 1 × 1 vial
RNA Dilution Buffer (RDB) 1 × 9mL
Quantity 10 Preps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.