Learn More
Invitrogen™ RELM alpha Monoclonal Antibody (226033)

Description
In direct ELISAs and Western blots, this antibody does not cross-react with recombinant mouse (rm) RELM beta or rmResistin. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | RELM alpha |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 226033 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | Retnla |
| Gene Accession No. | Q9EP95 |
| Gene Alias | 1810019L16Rik; cysteine-rich secreted protein A12-gamma; Cysteine-rich secreted protein FIZZ1; Fizz1; Fizz-1; FIZZ2; found in inflammatory zone 1; Himf; HXCP2; Hypoxia-induced mitogenic factor; parasite-induced macrophage novel gene 1 protein; Pmng1; RELMa; RELMalpha; RELM-alpha; RELMb; RELMbeta; RELM-beta; resistin like alpha; resistin-like alpha; Retnla; Xcp2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.