Learn More
RELM beta Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
$258.00 - $329.00
Specifications
| Antigen | RELM beta |
|---|---|
| Concentration | 0.1-1.0 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
500P21550UG
|
PeproTech
500P21550UG |
50 μg |
Each for $258.00
|
|
|||||
500P215100
|
PeproTech
500P215100UG |
100 μg |
Each for $329.00
|
|
|||||
Description
AA Sequence of recombinant protein: MQCSFESLVD QRIKEALSRQ EPKTISCTSV TSSGRLASCP AGMVVTGCAC GYGCGSWDIR NGNTCHCQCS VMDWASARCC RMA. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine RELMbeta. Anti-Murine RELMbeta-specific antibody was purified by affinity chromatography employing an immobilized Murine RELMbeta matrix. Sandwich ELISA: To detect Murine RELMbeta by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Murine RELMbeta (500-P215Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Murine RELMbeta. Western Blot: To detect Murine RELMbeta by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. when used in conjunction with compatible secondary reagents, the detection limit for Recombinant Murine RELMbeta is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Adipose tissue is an energy reserve that is strictly regulated in animals. Adipose cells produce and secrete numerous physiologically important proteins, such as lipoprotein lipase (LPL), leptin, adipocyte complement related protein of 30 kDa (Acrp30), perilipin, and resistin. Resistin, also known as Adipocyte Secreted Factor (ADSF) and FIZZ3, is a cytokine that is specifically secreted by adipocytes. While the function of resistin is not well understood, it appears to have key paracrine roles and has a suggested role in type 2 diabetes. Studies show that in obese, diabetic mice the levels of serum resistin are increased, which has an antagonistic effect on insulin. Additional studies have shown that resistin causes insulin resistance and glucose intolerance in mice. Resistin gene expression requires C/EBP alpha, which is necessary and sufficient for expression. Studies show that the coactivators CBP and p300, which have been shown to display histone acetyltransferase (HAT) activity, abundantly acetylate the histones at the resistin promoter suggesting their role in activating chromatin for resistin transcription. Numerous PPAR gamma and RXR ligands have been shown to downregulate resistin expression in adipocytes.Specifications
| RELM beta | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| Q99P86 | |
| 57263 | |
| E.coli-derived Recombinant Murine RELMbeta. | |
| Antigen affinity chromatography | |
| Retnlb |
| 0.1-1.0 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| PBS with no preservative | |
| 9030012B21Rik; C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich protein precursor 2; CCRG; Colon and small intestine-specific cysteine-rich protein; colon carcinoma-related gene protein; cysteine-rich secreted A12-alpha-like protein 1; cysteine-rich secreted protein A12-alpha-like 1; Cysteine-rich secreted protein A12-beta; cysteine-rich secreted protein FIZZ2; FIZZ1; Fizz2; found in inflammatory zone 1; found in inflammatory zone 2; HXCP2; Relmb; RELMbeta; RELM-beta; resistin like beta; resistin-like beta; resistin-like molecule beta; RETNL2; RETNLB; RSNB; UNQ408/PRO770; XCP2; Xcp3 | |
| Retnlb | |
| Primary | |
| -20°C |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.