Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RELM beta Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572118
Description
Adipose tissue is an energy reserve that is strictly regulated in animals. Adipose cells produce and secrete numerous physiologically important proteins, such as lipoprotein lipase (LPL), leptin, adipocyte complement related protein of 30 kDa (Acrp30), perilipin, and resistin. Resistin, also known as Adipocyte Secreted Factor (ADSF) and FIZZ3, is a cytokine that is specifically secreted by adipocytes. While the function of resistin is not well understood, it appears to have key paracrine roles and has a suggested role in type 2 diabetes. Studies show that in obese, diabetic mice the levels of serum resistin are increased, which has an antagonistic effect on insulin. Additional studies have shown that resistin causes insulin resistance and glucose intolerance in mice. Resistin gene expression requires C/EBP alpha, which is necessary and sufficient for expression. Studies show that the coactivators CBP and p300, which have been shown to display histone acetyltransferase (HAT) activity, abundantly acetylate the histones at the resistin promoter suggesting their role in activating chromatin for resistin transcription. Numerous PPAR gamma and RXR ligands have been shown to downregulate resistin expression in adipocytes.
Specifications
RELM beta | |
Polyclonal | |
Unconjugated | |
RETNLB | |
RETNLB ; resistin like beta ; FIZZ1; FIZZ2; HXCP2; RELM-beta; RELMb; RELMbeta; XCP2; C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich protein precursor 2; colon and small intestine-specific cysteine-rich protein; cysteine-rich secreted A12-alpha-like protein 1; found in inflammatory zone 1 | |
Rabbit | |
Protein A | |
RUO | |
84666 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.75 mg/mL | |
PBS with 0.09% sodium azide | |
Q9BQ08 | |
RETNLB | |
KLH conjugated synthetic peptide between 27-52 amino acids from the Central region of human RETNLB | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction