Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RELT/TNFRSF19L Antibody (111), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289555MFV610
Description
RELT/TNFRSF19L Monoclonal antibody specifically detects RELT/TNFRSF19L in Human samples. It is validated for ELISA, ELISASpecifications
RELT/TNFRSF19L | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Sandwich ELISA | |
111 | |
ELISA, Sandwich ELISA Capture | |
Receptor expressed in lymphoid tissues, RELT tumor necrosis factor receptor, TNFRSF19LFLJ14993, tumor necrosis factor receptor superfamily member 19L, tumor necrosis factor receptor superfamily, member 19-like | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human RELT/TNFRSF19L (Accession#: NP_116260.2; Met1-Ala160). | |
0.1 mL | |
Cancer | |
84957 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction