Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ REM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562055
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562055 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562055 Supplier Invitrogen™ Supplier No. PA562055
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QEAKTPLHRR ASTPLPLSPR GHQPGRLSTV PSTQSQHPRL GQSASLNPPT QKPSPAPDDW SSESSDSEGS WEALYRVVLL GD Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 87%.

The protein encoded by this gene is a GTPase and member of the RAS-like GTP-binding protein family. The encoded protein is expressed in endothelial cells, where it promotes reorganization of the actin cytoskeleton and morphological changes in the cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen REM1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene REM1
Gene Accession No. O75628
Gene Alias E030011C07Rik; GD:REM; GEM; GES; GTPase-regulating endothelial cell sprouting; GTP-binding protein REM 1; horizin; rad and gem related GTP binding protein 1; rad and Gem-like GTP-binding protein 1; Ras; RAS (RAD and GEM)-like GTP-binding 1; RAS-like GTP binding protein Rem; RAS-like GTP-binding protein REM; Rem; Rem1; RRAD and GEM like GTPase 1; SI:zK76P14.3; similar to RAS-like GTP-binding protein REM; zgc:56144
Gene Symbols REM1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human REM1. Recombinant protein control fragment (Product #RP-101133).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28954
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.