Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ REM2 Monoclonal Antibody (B4-D6)
GREENER_CHOICE

Catalog No. PIMA532902
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532902 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532902 Supplier Invitrogen™ Supplier No. MA532902
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

REM2 Monoclonal Antibody for ICC/IF, Western Blot

Binds GTP saturably and exhibits a low intrinsic rate of GTP hydrolysis. Target protein is a Ras-related small GTP-binding protein that is found in high levels in neuronal tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen REM2
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone B4-D6
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene REM2
Gene Accession No. Q8IYK8, Q8VEL9
Gene Alias AW411893; GEM, REM, Kir family small G-protein; GTP-binding protein REM 2; GTP-binding protein REM2; rad and gem related GTP binding protein 2; rad and Gem-like GTP-binding protein 2; Rad and Gem-related 2 (rat homolog); RAS (RAD and GEM) like GTP binding 2; RAS (RAD and GEM)-like GTP binding 2; RAS-like GTP binding 2; Ras-related GTP-binding protein of the Rad/Gem/Kir family member 2; Ras-related GTP-binding protein of the Rad/Gem/Kir family, member 2; Rem2; RRAD and GEM like GTPase 2
Gene Symbols REM2
Host Species Mouse
Immunogen Recombinant protein within Human Rem2 aa 1-340.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140743, 161253
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.