Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ REM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571399
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571399 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571399 Supplier Invitrogen™ Supplier No. PA571399
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

REM2 Polyclonal Antibody for Western Blot

Binds GTP saturably and exhibits a low intrinsic rate of GTP hydrolysis. Target protein is a Ras-related small GTP-binding protein that is found in high levels in neuronal tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen REM2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene REM2
Gene Accession No. Q8IYK8
Gene Alias AW411893; GEM, REM, Kir family small G-protein; GTP-binding protein REM 2; GTP-binding protein REM2; rad and gem related GTP binding protein 2; rad and Gem-like GTP-binding protein 2; Rad and Gem-related 2 (rat homolog); RAS (RAD and GEM) like GTP binding 2; RAS (RAD and GEM)-like GTP binding 2; RAS-like GTP binding 2; Ras-related GTP-binding protein of the Rad/Gem/Kir family member 2; Ras-related GTP-binding protein of the Rad/Gem/Kir family, member 2; Rem2; RRAD and GEM like GTPase 2
Gene Symbols REM2
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human REM2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161253
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.