Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
REM2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | REM2 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
REM2 Polyclonal specifically detects REM2 in Human samples. It is validated for Western Blot.Specifications
REM2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
CD300LE, FLJ38964, GTP-binding protein REM 2, ICLM2, Rad and Gem-like GTP-binding protein 2, Rad and Gem-related 2 (rat homolog), RAS (RAD and GEM) like GTP binding 2, RAS (RAD and GEM)-like GTP binding 2 | |
The immunogen is a synthetic peptide directed towards the middle region of human REM2 (NP_775798.2). Peptide sequence FEGAVRQIRLRRGRNHAGGQRPDPGSPEGPAPPARRESLTKKAKRFLANL | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
161253 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title