Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Remdesivir
GREENER_CHOICESDP

Catalog No. 722650 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-265-0 50 mg
72-261-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 72-265-0 Supplier Tocris Bioscience™ Supplier No. 7226/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Viral RNA-dependent RNA polymerase (RdRP) inhibitor; broad spectrum antiviral nucelotide prodrug

Remdesivir is a viral RNA-dependent RNA polymerase inhibitor (RdRp) and broad spectrum antiviral nucleotide prodrug. Metabolized to active form, GS 441524 (Cat. No. 7227). Exhibits activity against a range of RNA viruses in vitro (EC50 values are 0.019, 0.07, 0.07 - 0.34, 0.086 - 0.14, and 23.15 μM against RSV, SARS-CoV, MERS-CoV, EBOV and SARS-CoV-2, respectively). Exhibits antiviral efficacy in animal models of MERS-CoV and EBOV infection.

Specifications

Chemical Name or Material 2-Ethylbutyl (2 S)-2-[[-[[(2 R,3 S,4 R,5 R)-5-(4-aminopyrrolo(2,1-f)(1,2,4)triazin-7-yl)-5-cyano-3,4-dihydroxytetrahydrofuran-2-yl]methoxy]phenoxyphosphoryl]amino]propanoate
CAS 1809249-37-3
Quantity 50 mg
Target RNA/DNA Polymerase Inhibitors
Molecular Formula C27H35N6O8P
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.