Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Remel™ 2X Lauryl Tryptose Broth (LST Broth) (Lauryl Sulfate Tryptose)
Click to view available options
Quantity:
100 x 10 mL
Description
Thermo Scientific™ Remel Lauryl Tryptose Broth, 2X (LST Broth) (Lauryl Sulfate Tryptose) is a Standard Methods medium for detection of coliform bacteria in water, wastewater, and foods.
Specifications
Specifications
| Certifications/Compliance | Industrial Reference: AOAC, BAM, COMPF, EPA, SMWW, USDA. |
| Description | Lauryl Tryptose Broth, 2X |
| Format | 20mm x 150mm Durham Tube |
| Product Type | Microbiology Media |
| Quantity | 100 x 10 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction