Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Skim Milk Medium, 2X

Catalog No. p-4523922
Click to view available options
Format:
15mm x 103mm Tube
This item is not returnable. View return policy
This item is not returnable. View return policy

Differentiate microorganisms based on coagulation and proteolysis of casein using 2X Skim Milk Medium.

Differentiate microorganisms based on coagulation and proteolysis of casein with Thermo Scientific™ Remel™ 2X Skim Milk Medium. Skim milk is used as a substrate in media to identify psychrotropic bacteria which are commonly associated with milk products1. Such bacteria produce proteolytic enzymes that often remain active after treatment with ultra-high temperature sterilization2. Skim milk is also included in clinical laboratories to cultivate and identify organisms such as Mycobacterium tuberculosis, Cornybacterium diphtheriae, Clostridium butyricum3.

  • Also used as additive in microbiological media
TRUSTED_SUSTAINABILITY

Specifications

Description Skim Milk Medium 2X
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 20 x 5 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.