Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Remel™ A-7 Agar
Click to view available options
Format:
15mm x 60mm Plate
Media Type:
A-7 Agar
Description
Thermo Scientific™ Remel A-7 Agar is used for isolation and differentiation of Ureaplasma and some species of Mycoplasma.
Specifications
Specifications
| Format | 15mm x 60mm Plate |
| Media Type | A-7 Agar |
| Product Type | Agar |
| Quantity | 10/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction