Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, E. coli Polyvalent
Catalog No. p-4528205
Click to view available options
Description:
E. coli Polyvalent 2
E. coli Polyvalent 3
E. coli Polyvalent 4
This item is not returnable. View return policy
Catalog No. R30955101
Supplier: Thermo Scientific™ R30955101
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Used for serological identification of E. coli by slide and/or tube agglutination tests.
Remel Agglutinating Sera, E. coli Polyvalent is used for serological identification of E. coli by slide and/or tube agglutination tests.
TRUSTED_SUSTAINABILITY
Specifications
Description E. coli Polyvalent 4
Detectable Analytes E. coli
Type Agglutinating Sera
Packaging Type 2 mL/Vial
Quantity 2 mL
Specificity types O44, O112, O124, O142
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.