Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Neisseria meningitidis Agglutinating Sera

Catalog No. r30166601
Click to view available options
Quantity:
2 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Meningococcus Agglutinating Sera are used in the qualitative serological identification of Neisseria meningitidis Groups A, B, C, D, X, Y, Z, W135, A-D and X-Z + W135.

Perform serological identification of Neisseria meningitidis Groups A, B, C, D, X, Y, Z and W135 for epidemiological and diagnostic purposes using Thermo Scientific™ Remel™ Meningococcus Agglutinating Sera. Group specificity is based on the identity of capsular polysaccharides and strains possessing these materials are specifically agglutinated by the homologous antiserum. In cases of bacterial meningitis it is important in to obtain a presumptive identification of the infecting agent as early as possible, so that adequate antimicrobial therapy can be started.

Serological tests are based on the fact that antibodies in serum, produced in response to exposure to bacterial antigens, will agglutinate with bacteria carrying homologous antigens. Homologous cultures should be used for positive control organisms.

  • Ready to use - Prepared reagents
  • Easy to store - Store at 2-8°C
  • Polyvalent sera are for slide use only.
  • Easy to interpret -
    • Test suspension - Strong and clearly visible agglutination within one minute.
    • Control suspension - No visible agglutination.

Meningococcus Agglutinating Serum is produced in rabbits and preserved with 0.5% phenol. Each dropper bottle contains 2 mL liquid and is ready for use.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Neisseria meningitidis Polyvalent Groups A - D
CE Marker Yes
Detectable Analytes N. meningitidis
Type Neisseria Meningitidis Test Kit
Material Polyvalent
Organism Group Polyvalent Groups A-D
Packaging Type 2 mL/Vial
Quantity 2 mL
Target Organism Class Neisseria Meningitidis
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.