Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella H
Catalog No. r30160401
Click to view available options
Description:
Polyvalent, Phase 2
Polyvalent, Phases 1 and 2
Rapid Diagnostic 3 (d, E, G, k)
Salmonella H (a)
Salmonella H (b)
Salmonella H (c )
Salmonella H (d)
Salmonella H (i)
Salmonella H (r )
This item is not returnable. View return policy
This item is not returnable. View return policy
Used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
Remel Agglutinating Sera, Salmonella H is used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
  • Select sera also for use as positive controls for stained Salmonella suspensions
TRUSTED_SUSTAINABILITY
Specifications
Control Sets Positive
Description Salmonella H (c )
CE Marker Yes
Detectable Analytes Salmonella H Antigen
Type Agglutinating Sera
Packaging Type 2 mL/Vial
Quantity 2 mL
Target Organism Class Salmonella
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.