Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Alsevers Sheep Blood

Catalog No. r54048
Click to view available options
Quantity:
50 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Be confident that your animal blood meets the highest standards possible by using Remel Alsevers Sheep Blood, sourced from our own technologically-advanced sheep farm.

Be confident that your animal blood meets the highest standards possible by using Remel Alsevers Sheep Blood, sourced from our own technologically-advanced sheep farm.

TRUSTED_SUSTAINABILITY

Specifications

Species Sheep
Form Liquid
Additive Alsevers
Product Type Whole Blood
Quantity 50 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.