Learn More
Thermo Scientific™ Remel™ Anaerobic Blood Agar, Monoplate
Catalog No. R01039
Click to view available options
Quantity:
10/Pk.
100/Pk.
15/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy
Isolate and cultivate anaerobic organisms, including fastidious strains with this ready-to-use medium.
Isolate and cultivate anaerobic organisms, including fastidious strains with Thermo Scientific™ Remel™ Anaerobic Blood Agar ready prepared plates. Anaerobic Blood Agar serves as a nutritious medium supporting good growth and typical pigmentation of fastidious and slow growing anaerobes, as well as other anaerobes of clinical importance.
Anaerobe Blood Agar contains peptones which supply nitrogenous substances and amino acids necessary for the growth of anaerobic bacteria. Yeast extract provides B-complex vitamins and serves as a growth enhancer. Hemin, vitamin K, and sheep blood stimulate the growth of anaerobes. Sodium chloride is a source of essential electrolytes and maintains osmotic equilibrium.
TRUSTED_SUSTAINABILITY
Specifications
Description Anaerobic Blood Agar
Format 10mm x 100mm Spacesaver Plate
Product Type Agar
Quantity 15/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.