Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Anaerobic KV Blood Agar (CDC formulation) with kanamycin and vancomycin

Catalog No. p-4523664
Click to view available options
Quantity:
10/Pk.
100/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

Inhibit gram-positive microorganisms and facultative anaerobic bacteria and select for gram-negative bacilli with this selective, ready-to-use solid medium.

Inhibit gram-positive microorganisms and facultative anaerobic bacteria and select for gram-negative bacilli with Thermo Scientific™ Anaerobic KV Blood Agar [The Centers for Disease Control and Prevention (CDC) formulation] with kanamycin and vancomycin.

Formulated by V.R. Dowell and T.M Hawkins at the CDC in Atlanta Georgia1, Anaerobic Blood Agar (CDC) supports the growth and typical pigmentation of fastidious, slow-growing and other clinically important anaerobes.

Isolate and cultivate anaerobic organisms, including fastidious strains using the solid medium Remel Anaerobic KV Blood Agar (CDC formulation) with kanamycin and vancomycin.

  • Selective: Kanamycin and vancomycin are antibiotics which inhibit gram-positive organisms and facultative anaerobic bacteria and select for gram-negative bacilli.
  • Ready-to-use: Convenience of prepared media.

Anaerobic Blood Agar (CDC) base contains peptones that supply nitrogenous substances and amino acids necessary for the growth of anaerobic bacteria. Yeast extract provides B-complex vitamins and serves as a growth enhancer. Hemin, vitamin K and sheep blood stimulate the growth of anaerobes. Sodium chloride is a source of essential electrolytes and maintains osmotic equilibrium. Kanamycin and vancomycin are antibiotics which inhibit gram-positive organisms and facultative anaerobic bacteria and select for gram-negative bacilli.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Anaerobic KV Blood Agar
Format 12mm x 85mm Monoplate
Product Type Agar
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.