Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Anaerobic (CDC) Neomycin Blood Agar
Click to view available options
Format:
10mm x 100mm Spacesaver Plate
Description
Support the growth and typical pigmentation of fastidious, slow-growing and other clinically-important anaerobes with Thermo Scientific™ Remel™ Anaerobic (CDC) Neomycin Blood Agar [The Centers for Disease Control and Prevention (CDC) formulation]1
Adding neomycin facilitates the isolation of anaerobic organisms, such as Bacteriodes and Clostridium spp. and inhibits enteric gram-negative bacilli.
- Neomycin inhibits gram-negative Enterobacteriaceae

Specifications
Specifications
Description | Anaerobic Neomycin Blood Agar |
Format | 10mm x 100mm Spacesaver Plate |
Product Type | Agar |
Quantity | 15/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction