Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Anaerobic PEA Blood Agar (Anaerobic Phenylethyl Alcohol Agar) with 5% Sheep Blood

Catalog No. p-4523671
Click to view available options
Quantity:
10/Pk.
100/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

Isolate and cultivate anaerobic organisms, including fastidious strains with this selective, ready-to-use medium.

Isolate and cultivate anaerobic organisms, including fastidious strains with Thermo Scientific™ Remel™ Anaerobic Blood Agar ready prepared plates. Anaerobic Blood Agar serves as a nutritious medium supporting good growth and typical pigmentation of fastidious and slow growing anaerobes, as well as other anaerobes of clinical importance. Selective agents like kanamycin, vancomycin, neomycin and paramomycin make the medium selective for fastidious, anaerobic bacteria from mixed populations by inhibiting facultative and aerobic gram-negative organisms.

Use Anaerobic Blood Agar for isolation and cultivation of anaerobic organisms including the fastidious strains.

  • Selective: the addition of phenylethyl alcohol inhibits facultative anaerobic gram-negative bacilli.
  • Ready-to-use: convenience of prepared media.

Anaerobe Blood Agar contains peptones which supply nitrogenous substances and amino acids necessary for the growth of anaerobic bacteria. Yeast extract provides B-complex vitamins and serves as a growth enhancer. Hemin, vitamin K, and sheep blood stimulate the growth of anaerobes. Sodium chloride is a source of essential electrolytes and maintains osmotic equilibrium.

Not all products are available for sale in all territories. Please inquire.
 

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Anaerobic PEA Blood Agar
Format 12mm x 85mm Monoplate
Product Type Agar
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.