Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Anaerobic Reducible Blood Agar with colistin and nalidixic acid (CNA)
Click to view available options
Quantity:
10/Pk.
100/Pk.
Description
Cultivate and isolate fastidious anaerobic bacteria with Thermo Scientific™ Remel™ Anaerobic Reducible Blood Agar.
The addition of cysteine, palladium chloride, and dithiothreitol makes the medium reducible. Reducible medium improves the recovery of anaerobes by lowering the oxidation – reduction potential of the medium. Kanamycin and vancomycin are added as selective agents for recovery of Gram-negative anaerobes, especially Bacteriodes spp
- Colistin and nalidixic acid inhibit gram-negative bacilli
Specifications
Specifications
| Description | Anaerobic Reducible; CNA Agar Base |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar |
| Quantity | 100/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction