Learn More
Thermo Scientific™ Anaerobic Reducible Blood Agar with colistin and nalidixic acid (CNA)
Catalog No. r01063
Click to view available options
Quantity:
10/Pk.
100/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy
Cultivate and isolate fastidious anaerobic bacteria with this selective, ready-to-use solid medium.
Cultivate and isolate fastidious anaerobic bacteria with Thermo Scientific™ Remel™ Anaerobic Reducible Blood Agar.
The addition of cysteine, palladium chloride, and dithiothreitol makes the medium reducible. Reducible medium improves the recovery of anaerobes by lowering the oxidation – reduction potential of the medium. Kanamycin and vancomycin are added as selective agents for recovery of Gram-negative anaerobes, especially Bacteriodes spp
  • Colistin and nalidixic acid inhibit gram-negative bacilli
TRUSTED_SUSTAINABILITY
Specifications
Description Anaerobic Reducible; CNA Agar Base
Format 12mm x 85mm Monoplate
Product Type Agar
Quantity 100/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.