Learn More
Thermo Scientific™ Remel™ BactiDrop Acridine Orange
Description
Detect microorganisms from clinical specimens and blood cultures using Thermo Scientific™ Remel™ BactiDrop™ Acridine Orange fluorescent stain. Kronvall and Myhre demonstrated that acridine orange stain detected microorganisms in direct smears1 and when buffered at a low pH produced differential staining of bacteria. It was evaluated for the detection of microorganisms in blood cultures2 and was found to be a rapid, inexpensive alternative to blind subcultures and more sensitive than Gram stains. Subsequent studies showed acridine orange stain to be more sensitive than the Gram stain3.
Specifications
Specifications
| Type | Acridine Orange Stain |
| Quantity | 50 Pk. |
| Volume (Metric) | 0.75 mL/Ampule |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.