Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ BactiDrop™ Desoxycholate
GSA_VA

Catalog No. R21508 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 x 0.75 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R21508 50 x 0.75 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R21508 Supplier Thermo Scientific™ Supplier No. R21508
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Differentiate between bile soluble Streptococcus pneumoniae and other insoluble alpha-hemolytic streptococci using BactiDrop™ Desoxycholate (10%).

Differentiate between bile soluble Streptococcus pneumoniae and other insoluble alpha-hemolytic streptococci using Thermo Scientific™ Remel™ BactiDrop™ Desoxycholate (10%). In 1900, Neufeld used a bile salt sodium desoxycholate to detect autolysis, that differentiated bile-soluble S. pneumoniae from bile-insoluble alpha-hemolytic streptococci1. Evans employed 10% bile to lyse broth cultures of pneumococci2. Hawn and Beebe evaluated the effectiveness of applying bile salts directly to a blood agar plate culture of pneumococci3.

Use Bactidrop Desoxychocolate to differentiate between bile soluble Streptococcus pneumoniae and other insoluble alpha-hemolytic streptococci.Ready-to-use: Liquid reagentEasy to interpret:Spot Test Positive Test - Colony disintegration within 30 minutesNegative Test - Colony remains intact within 30 minutesTube Test Positive Test - Clearing of Test suspension within 3 hours; Control suspension remains turbidNegative Test - Test and Control suspensions remain turbid within 3 hoursThe bile solubility test is based on the observation that when 10% desoxycholate is applied S. pneumoniae cells lyse, while other streptococci and gram-positive cocci do not. Bile salts cause a cell membrane derangment and also lower surface tension at the medium-membrane interface4,5.S. pneumoniae possesses an autocatalytic enzyme that lyses the organism’s own cell wall during cell division. This autolytic intracellular enzyme causes the organism to undergo rapid autolysis when cultivated on artificial medium6. Goebel and Avery stated that the action of bile salts is independent of the autolytic enzyme7. Dubos and Hirsch found that bile salts inactivate the naturally occurring autolytic enzyme either by the alteration or the removal of a normal autolytic inhibitor from the pneumococcal cells8.This reagent contains 10% w/v desoxycholic acid sodium, ethanol, sodium Merthiolate and demineralized water.Not all products are available for sale in all territories. Please inquire.Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 20°C to 25°C
Description BactiDrop Desoxycholate
CE Marker Yes
Packaging Type 50/Pk.
Quantity 50 x 0.75 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.