Learn More
Thermo Scientific™ Brain Heart Infusion (BHI) Agar
Catalog No. p-4524007
Click to view available options
Quantity:
100/Pk.
20/Pk.
Format:
15mm x 103mm Tube
20mm x 113mm Tube - Slant
20mm x 150mm Pour Tube
This item is not returnable. View return policy
This item is not returnable. View return policy
Isolate and cultivate a wide variety of micro-organisms including pathogenic fungi this ready-to-use medium recommended by AOAC and APHA.
Isolate and cultivate a wide variety of microorganisms including pathogenic fungi with Thermo Scientific™ Remel™ Brain Heart Infusion (BHI) Agar. Due to the addition of 5-10% sheep blood, BHI agar can be used for the isolation of dimorphic fungi which may grow poorly on non-enriched media. BHI Agar is recommended by the Association of Official Analytical Chemists (AOAC) and the American Public Health Association (APHA) 1, 2
Use Heart Infusion Agar for the cultivation of a wide variety of microorganisms including pathogenic fungi.
  • Recommended by AOAC and APHA.
  • Ready-to-use convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description BHI Agar (Brain Heart Infusion Agar)
Format 15mm x 103mm Tube
Product Type Agar
Quantity 20/Pk.
REF R112025 not intended for IVD use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.