Learn More
Thermo Scientific™ Remel™ BHI Agar w/Chloramphenicol, Cycloheximide
Catalog No. p-4523831
Click to view available options
Quantity:
100/Pk.
20/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy
Thermo Scientific™ Remel BHI Agar with Chloramphenicol and Cycloheximide is for the selective isolation of fungi and yeast.
Thermo Scientific™ Remel BHI Agar with Chloramphenicol and Cycloheximide is for the selective isolation of fungi and yeast.
Chloramphenicol inhibits some gram-positive and gram-negative bacteria. Cycloheximide inhibits rapid growing saprophytic fungi.
TRUSTED_SUSTAINABILITY
Specifications
Description BHI Agar w/Chloramphenicol, Cycloheximide
Format 20mm x 113mm Tube - Slant
Product Type Agar
Quantity 100/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.