Learn More
Thermo Scientific™ Remel™ BHI Agar w/Sheep Blood, Chloramphenicol, Gentamicin, Cycloheximide
Catalog No. p-4522889
Click to view available options
Quantity:
10/Pk.
100/Pk.
20/Pk.
Format:
F - Slants
Monoplate Deep Pour
Packaging Type:
10/Pk.
100/Pk.
20/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy
Thermo Scientific™ Remel BHI Agar w/Sheep Blood, Chloramphenicol, Gentamicin, Cycloheximide is used for selective isolation of fastidious fungi and yeast.
Thermo Scientific™ Remel BHI Agar w/Sheep Blood, Chloramphenicol, Gentamicin, Cycloheximide is used for selective isolation of fastidious fungi and yeast.
  • Chloramphenicol inhibits some gram-positive and gram-negative microorganisms
  • Gentamicin inhibits some gram-negative bacteria
  • Cycloheximide inhibits rapid growing saprophytic fungi
TRUSTED_SUSTAINABILITY
Specifications
Description BHI Agar w/Sheep Blood, Chloramphenicol, Gentamicin, Cycloheximide
Format Monoplate Deep Pour
Product Type Agar
Packaging Type 10/Pk.
Quantity 10/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.