Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BHI Agar with Sheep Blood

Catalog No. r08066
Click to view available options
Quantity:
10/Pk.
100/Pk.
20/Pk.
Format:
20mm x 113mm Tube
Monoplate Deep Pour
This item is not returnable. View return policy
This item is not returnable. View return policy

Cultivate a wide variety of microorganisms including bacteria, yeasts, and molds with this nutritious medium.

Cultivate a wide variety of microorganisms including bacteria, yeasts, and molds with nutritious Thermo Scientific™ Remel™ BHI Agar with Sheep Blood. Due to the addition of Sheep blood, the BHI Agar can be used for the isolation of dimorphic fungi which may grow poorly on non-enriched media.

Use BHI Agar with Sheep Blood for the cultivation of a wide variety of micro-organisms including pathogenic fungi.

  • Highly Nutritious: medium recommended for the enrichment of fastidious organisms.
  • Ready-to-use: convenience of prepared media.

BHI Agar w/Sheep Blood contains brain heart infusion, peptone, and dextrose which provide nitrogen, carbon, sulfur, vitamins and a carbohydrate source. Sheep blood provides essential growth factors for fastidious fungi. Yeast extracts are incorporated in the medium to serve as a growth stimulant.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Description BHI Agar w/Sheep Blood
Format 20mm x 113mm Tube
Product Type Agar
Quantity 20/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.