Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BHI Agar

Catalog No. R01130
Click to view available options
Format:
12mm x 85mm Monoplate
15mm x 103mm Tube
Monoplate Deep Pour
This item is not returnable. View return policy
This item is not returnable. View return policy

Cultivate a wide variety of microorganisms including bacteria, yeasts, and molds with the nutritious, ready-to-use base medium.

Cultivate a wide variety of microorganisms including bacteria, yeasts, and molds with the nutritious base medium Thermo Scientific™ Remel™ Blood Heart Infusion (BHI) Agar with Sheep Blood1.

Use BHI Agar for the cultivation of a wide variety of microorganisms, including pathogenic fungi.

  • Highly Nutritious: medium recommended for the enrichment of fastidious organisms.
  • Ready-to-use: the convenience of prepared media.

BHI Agar contains brain heart infusion, peptone, and dextrose, which provide nitrogen, carbon, sulfur, vitamins and a carbohydrate source. Sheep blood provides essential growth factors for fastidious fungi. Yeast extracts are incorporated in the medium to serve as a growth stimulant. BHI Agar can also be used for: Cryptococcus neoformans ATCC® 14116, Escherichia coli ATCC® 25922 and Staphylococcus aureus ATCC® 25923.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: BAM, COMPF, EPA, IDF, SMD.
Description BHI Agar (Brain Heart Infusion Agar)
Format 12mm x 85mm Monoplate
Product Type Agar
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.