Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ BHI Broth (Brain Heart Infusion Broth)

Catalog No. R14101
Click to view available options
Quantity:
20 x 2 mL
20 x 5 mL
100 x 0.5 mL
100 x 2 mL
100 x 5 mL
20 x 10 mL
10 x 500 mL
Format:
15mm x 103mm Tube
15mm x 103mm Tube - Filtered
20mm x 113mm Tube
Borosilicate Bottle

Thermo Scientific™ Remel BHI Broth (Brain Heart Infusion Broth) is for the cultivation of microorganisms.

Thermo Scientific™ Remel BHI Broth (Brain Heart Infusion Broth) is for the cultivation of microorganisms.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: BAM, SMD, SMWW, USDA.
Description BHI Broth
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 100 x 0.5 mL

REF R14101 for use with MicroScan™ System. REF R112022 not intended for IVD use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.