Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Remel™ BHI Broth w/6.5% NaCl
Click to view available options
Quantity:
20 x 4.5 mL
100 x 4.5 mL
Description
Thermo Scientific™ Remel BHI Broth with 6.5% NaCl (Brain Heart Infusion Broth) is for the differentiation of enterococci from nonenterococcal group D streptococci.

Specifications
Specifications
Description | BHI Broth w/6.5% NaCl |
Format | 15mm x 103mm Tube |
Product Type | Microbiology Media |
Quantity | 20 x 4.5 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction