Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ BHI Broth w/Fildes Enrichment

Catalog No. p-4523782
Click to view available options
Quantity:
20 x 5 mL
100 x 5 mL
This item is not returnable. View return policy

Catalog No. R060280

Supplier: Thermo Scientific™ R060280

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Thermo Scientific™ Remel BHI Broth w/Fildes Enrichment (Brain Heart Infusion Broth) is for the cultivation of Haemophilus spp.

Thermo Scientific™ Remel BHI Broth w/Fildes Enrichment (Brain Heart Infusion Broth) is for the cultivation of Haemophilus spp.

TRUSTED_SUSTAINABILITY

Specifications

Description BHI Broth w/Fildes Enrichment
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 100 x 5 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.