Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ BHI Broth w/Hemin, Vitamin K

Catalog No. r060276
Click to view available options
Quantity:
20 x 5 mL
100 x 5 mL
Format:
15mm x 103mm Tube
This item is not returnable. View return policy

Catalog No. R060276

Supplier: Thermo Scientific™ R060276

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Thermo Scientific™ Remel BHI Broth with Hemin, Vitamin K (Brain Heart Infusion Broth) is for the cultivation of aerobic and anaerobic microorganisms.

Thermo Scientific™ Remel BHI Broth with Hemin, Vitamin K (Brain Heart Infusion Broth) is for the cultivation of aerobic and anaerobic microorganisms.

TRUSTED_SUSTAINABILITY

Specifications

Description BHI Broth w/Hemin; Vitamin K
Format 15mm x 103mm Tube
Product Type Microbiology Media
Packaging Type 100/Case
Quantity 100 x 5 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.