Learn More
Thermo Scientific™ Remel™ Biplates: Anaerobic Reducible through Levine Agars
Catalog No. p-4522972
Click to view available options
Quantity:
10/Pk.
100/Pk.
Media Type:
Chocolate /Thayer Martin Agar
Columbia CNA/MacConkey Agar
This item is not returnable. View return policy
Catalog No. R02058
Supplier: Thermo Scientific™ R02058
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Perform screening and isolation in the same 12 x 85mm dish, divided into two sections to hold either the same media on both sides or two media, one on each side.
Perform screening and isolation in the same 12 x 85mm dish using Thermo Scientific™ Remel™ Biplates, divided into two sections to hold either the same media on both sides or two media, one on each side.
TRUSTED_SUSTAINABILITY
Specifications
Format Biplate
Media Type Chocolate /Thayer Martin Agar
Product Type Agar
Quantity 10/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.