Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Remel™ Birdseed Agar
Click to view available options
Format:
20mm x 150mm Pour Tube
Monoplate
Description
Thermo Scientific™ Remel Birdseed Agar is for the presumptive identification of Cryptococcus neoformans.
Specifications
Specifications
| Description | Birdseed Agar |
| Format | Monoplate |
| Product Type | Agar |
| Quantity | 10/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction