Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Blood Agar with Ampicillin (with 5% Sheep Blood)

Catalog No. R01226

Selectively isolate Aeromonas species from clinical specimens with this selective, ready-to-use medium.

Selectively isolate Aeromonas species from clinical specimens with Thermo Scientific™ Remel™ Blood Agar with Amipicillin. The addition of 5% sheep blood enriches the medium by providing the essential growth factors and provides for direct detection of hemolysis and oxidase. Ampicillin is a selective agent inhibitory to normal flora.

The genus Aeromonas contains ferementative, oxidase positive, Gram-negative bacilli  which are common inhabitants of aquatic ecosystem worldwide. When it is considered an uncommon pathogen, gastroenteritis caused by Aeromonas spp. is an increasingly recognised clinical entity, reported to occur on a global basis.

Use Blood Agar with Ampicillin (with 5% Sheep Blood) for selective isolation of Aeromonas species from clinical specimens.

  • Selective: ampicillin is selective for Aeromonas species.
  • Easy-to-read: Aeromonas species appear as gray and moist colonies with or without hemolyisis and consistently oxidase positive
  • Ready-to-use: convenience of prepared media.

 

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Description Blood Agar w/Ampicillin
Format 12mm x 85mm Monoplate
Product Type Agar
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.