Learn More
Thermo Scientific™ Remel™ Buffered CYE Agar
Catalog No. p-4523469
Click to view available options
Format:
12mm x 85mm Monoplate
This item is not returnable. View return policy
This item is not returnable. View return policy
Use for the cultivation and isolation of Legionella species.
Thermo Scientific™ Remel Buffered CYE Agar (Buffered Charcoal Yeast Extract) (BCYE) is used for the cultivation and isolation of Legionella species.
Ordering Information:
REF R09300 requires addition of BCYE supplement, REF R45005. Optional antimicrobial supplements are PAC, REF R45006, PAV, REF R45007.
TRUSTED_SUSTAINABILITY
Specifications
Description Buffered CYE Agar (BCYE)
Format 12mm x 85mm Monoplate
Product Type Agar
Quantity 10/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.