Learn More
Thermo Scientific™ Remel™ Buffered Peptone Water
Catalog No. R452672
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R452672 500 g
R452674 2.5 kg
R452676 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R452672 Supplier Thermo Scientific™ Supplier No. R452672
Thermo Scientific™ Remel Buffered Peptone Water is used for the non-selective pre-enrichment of Salmonella spp. from foods.
Thermo Scientific™ Remel Buffered Peptone Water is used for the non-selective pre-enrichment of Salmonella spp. from foods.
TRUSTED_SUSTAINABILITY
Specifications
Description Buffered Peptone Water
Form Powder
Product Type Peptone Water
Quantity 500 g
For Laboratory Use Only