Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Calgiswab™ Type 3 Swab: Swab with Plastic-shaft

Catalog No. R6015003
Click to view available options
Quantity:
100/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

Thermo Scientific™ Remel Calgiswab Swab system is designed for nasopharyngeal specimen collection.

Plastic-shaft swab for use with Thermo Scientific™ Remel Calgiswab Swab system. System designed for nasopharyngeal specimen collection.

Manufacturer: Puritan

Order Info

Swab only

TRUSTED_SUSTAINABILITY

Specifications

Product Type Remel Calgiswab
Format Swab
Description Plastic-shaft; 1/Envelope; 100/Pk.
For Use With (Application) Healthcare
Packaging 1/Envelope
Quantity 100/Pk.
Shaft Material Plastic
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.